Learn More
Description
Specifications
Specifications
| Concentration | 0.5mg/mL |
| Formulation | In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol |
| Gene ID (Entrez) | 384 |
| Molecular Weight (g/mol) | 38.3kDa |
| Purification Method | Protein |
| Quantity | 0.25mg |
| Source | Human |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MVHSVAVIGA PFSQGQKRKG VEHGPAAIRE AGLMKRLSSL GCHLKDFGDL SFTPVPKDDL YNNLIVNPRS VGLANQELAE VVSRAVSDGY SCVTLGGDHS LAIGTISGHA RHCPDLCVVW VDAHADINTP LTTSSGNLHG QPVSFLLREL QDKVPQLPGF SWIKPCISSA SIVYIGLRDV DPPEHFILKN YDIQYFSMRD IDRLGIQKVM ERTFDLLIGK RQRPIHLSFD IDAFDPTLAP ATFTPVVGGL TYREGMYIAE EIHNTGLLSA LDLVEVNPQL ATSEEEAKTT ANLAVDVIAS SFGQTREGGH IVYDQLPTPS SPDESENQAR VRI |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Common Name | ARG2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
