Learn More
CPC Scientific H-Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific VASP001A
Encompass
An antidiuretic peptide hormone that is released from the hypothalamus during periods of stress and implicated in a variety of physiological processes including diuresis, vasoregulation, thermoregulation and memory. AVP's primary functions include regulating water balance in animals (antidiuretic action) and arteriole contracting (vasopressor action), as it also affects the immune system.ONE-LETTER SEQUENCE: CYFQNCPRG-NH2 (C1&C6 bridge)MOLECULAR FORMULA: C46H65N15O12S2MOLECULAR WEIGHT:1084.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [113-79-1], SYNONYMS: AVP, Argipressin, (Phe3,Arg8)-Oxytocin, Leiormone, Arginine Antidiuretic HormoneRESEARCH AREA: HormonalREFERENCES: A.Lagumdzija et al., Regul. Pept., 121, 41 (2004); Y.Isobe et al., Brain Res., 918, 67 (2001); H.Hashimoto et al., Neuropeptides, 12, 199 (1988); J. Bell et al., Intl. J. Immunopharmacol., 14, 92 (1992)
Catalog No. 50-211-0605
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.