Learn More
Arbor Assays Arg8-Vasopressin (AVP) Chemiluminescent ELISA Kit (5 Plate)
SDP
Supplier:  Arbor Assays K049C5
Encompass_Preferred
The DetectX Arg8-Vasopressin (AVP) Chemiluminescent ELISA Kits quantitatively measure AVP present in serum, plasma and tissue culture media samples.
Catalog No. 50-227-8464
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.