Learn More
AdipoGen Argania Spinosa Kernel Oil (natural) (pure)
SDP
Supplier:  AdipoGen IAX700001L100
Encompass_Preferred
Argania Spinosa Kernel Oil (natural) (pure), AdipoGen Life Sciences. Chemical. CAS: 223747-87-3. Isolated from Argania spinosa. Argan oil is pressed from the kernels of the fruits from the argan tree (Argania spinosa (L.) Skeels; Sapotaceae) that is endemic to southwest Morocco. Cold pressed argan oil is characterized by high levels of linoleic and oleic acids, tocopherols (in particular ?-tocopherol), and minor compounds such as sterols, carotenoids, and squalene. The main fatty acids in these triglycerides, are oleic and linoleic acids (47% and 33%, respectively), as well as omega-6 fatty acids. Pre-clinical and clinical studies report hypolipidemic, hypocholesterolemic, hypoglycemic and antihypertensive effects of nutritional use of argan oil in addition to its well-documented anti-oxidant and moisturizing effects used for external cosmetic applications. Used for topical application, as food supplement and as solvent aid for lipophilic compounds. 223747-87-3
Catalog No. 50-117-6380
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.