Learn More
Apexbio Technology LLC Argatroban, 10mg. Cas: 74863-84-6 MFCD: MFCD00866789. Thrombin mediated fibrinogen cleavage inhibitor
SDP
Supplier:  Apexbio Technology LLC A836310
Encompass_Preferred
Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5127
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.