Learn More
Invitrogen™ Arginase 2 Monoclonal Antibody (OTI3G5)

Description
Arginase 2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | Arginase 2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI3G5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ARG2 |
| Gene Accession No. | O08691, O08701, P78540 |
| Gene Alias | AII; AII type II arginase; ARG2; arginase 2; arginase II; arginase type II; arginase, type II; arginase-2, mitochondrial; AU022422; kidney arginase; kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; nonhepatic arginase; non-hepatic arginase; type II arginase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.