Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Arginase, MP Biomedicals™

Catalog No. MP210027725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210027725 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210027725 Supplier MP Biomedicals™ Supplier No. 0210027725
Add to Cart
Add to Cart
Request a Quote

Converts arginine to ornithine. Arginase, MP Biomedicals is the major degradative enzyme for arginine. Arginase deficiency is associated with spasticity and motor dysfunction. It is most highly concentrated in the mammalian liver. It is also present in th

  • Arginase catalyzes the following reaction: Arginine + H2O → Ornithine + Urea
  • The enzyme participates in the Krebs-Henseleit urea cycle
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
pH Optimum pH: 10 (Mn2+ activated) (Lit.)
For Use With (Application) Arginase catalyzes the following reaction: Arginine + H2O → Ornithine + Urea. The enzyme participates in the Krebs-Henseleit urea cycle. It is most highly concentrated in mammalian liver. It is also present in the mammary glands.
Content And Storage Store at −20°C. Store Desiccated.
Quantity 25 kU
Molecular Weight (g/mol) 115,000–120,000 (Lit.)
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.