Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Essentials BioSafe Serological Pipettes

Catalog No. 03395179 Shop All Oakton Products
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
25 mL
Graduations:
0.1 mL
0.2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Graduations
03-395-179 10 mL 0.1 mL
03-395-178 5 mL 0.1 mL
03-395-180 25 mL 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 03-395-179 Supplier Cole-Parmer™ Essentials Supplier No. 1300253

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Filtered for sample integrity and user safety

  • Eliminate cross-contamination - unique barrier filter prevents over-aspiration
  • Polystyrene pipettes are sterile, individually wrapped, and disposable
  • Ideal for use with potentially hazardous or dangerous biological fluids
  • Use in place of filtered pipette tips
These polystyrene serological pipettes feature a special filter plug to lock in fluid and vapors if over aspiration occurs. They also protect electronic pipettors from contamination and cross-contamination of samples from over-aspiration - promoting user safety and sample integrity.

Specifications

Accuracy ±0.2 mL
Aspirator Plug Yes
Autoclavable Not Autoclavable
Calibrated Yes
Calibration To Deliver
Material Polystyrene
Sterility Sterile
For Use With (Application) Biological fluids
Graduated Yes
Graduations 0.1 mL
Quantity 200
Packaging Type Individually Wrapped
Color Clear
Volume (Metric) 10 mL
Length (Metric) 40.64 cm
Type Serological Pipet
Disposable Reusable
Show More Show Less

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.