Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ E-Vac Aspiration System

Catalog No. 10987031 Shop All Oakton Products
Change view
Click to view available options
Connection Type:
Quick-release Couplings
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
10-987-031 Quick-release Couplings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-987-031 Supplier Cole-Parmer™ Supplier No. 0439710

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

E-Vac 310 system includes 4L polypropylene bottle, lid with quick-release couplings, level detection sensor cable and HandEvac hand operator

Self-contained aspiration system-vacuum pump built into the base!

  • Keep clean-all components are autoclavable except the vacuum base
  • Protect your vacuum pump-included filter prevents liquid aspiration into the pump
  • Minimize down-time when collection bottle is full-additional bottles available
  • Be safe-plastic bottles won't shatter if dropped
  • Choose from model with a hose barb couplings cap or models with quick-release couplings cap and a collection bottle level sensor

The E-Vac aspiration system is designed to collect critical or dangerous liquids. Waste is collected in the bottle through vacuum, and pressure-sensitive button controls the right amount of vacuum for your application. All parts, except the stainless steel base containing the pump, are fully autoclavable. Two hydrophobic filters help prevent liquids from getting into the pump and pump housing in the event of an overflow.

The HandE-Vac hand operator, included with both systems options, is designed to fit the E-Vac systems or any vacuum device currently available on the market. Set vacuum by manual adjustment of the aperture control button. Ergonomic design allows for fatigue-free aspiration. The hand operator can also be used with Pasteur pipettes or multi-well plates and dishes, additional adapters required and sold separately.

Full bottles can be swapped out and autoclaved with minimum of effort and new bottles replaced within seconds. All parts, except the stainless steel base containing the pump, are fully autoclavable.

Certifications

CE approved, RoHS/WEEE compliant

Specifications

Max. Vacuum -250 to -650 mbar
Control Analog dial
Includes 4 L polypropylene bottle, lid with quick-release couplings, level detection sensor cable and HandE-Vac hand operator
Certifications/Compliance CE approved, RoHS/WEEE compliant
Electrical Requirements 100/240 VAC 50/60 Hz
Description E-Vac 310 Aspirating System- E-Vac base with 4L Polypropylene bottle and lid with quick release tubing connector and level detection sensor, 700mm tubing (EV331), 1800 mm tubing (EV332), Filter (EV015), HandEvac hand operator with single channel adapter (EV514)
Material Glass
Model EV310
Noise Level 50 dB ± 5 dB at 1 m
Plug Type Universal
Flow Rate 8 L/min.
Frequency 50/60 Hz
Pump Type Oilless Diaphragm Pump
Voltage 100/240 VAC
Connection Type Quick-release Couplings
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.