Learn More
Medchemexpress LLC ARGX-119 1mg | 1mg
Supplier: Medchemexpress LLC HYP9910661MG
ARGX-119 is a first-in-class humanized agonist monoclonal antibody specific for muscle-specific kinase (MuSK) ARGX-119 can be used for the study of neuromuscular diseases The isotype control for ARGX-119 can refer to Human IgG1 lambda1 Isotype Control (HY-P99992)[1]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.