Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101840 Supplier Invitrogen™ Supplier No. PA5101840
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARHGAP11A.

ARHGAP11A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP11A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGAP11A
Gene Accession No. Q6P4F7, Q80Y19
Gene Alias 6530401L14Rik; Arhgap11a; GAP (1-12); Kiaa0013; mKIAA0013; RGD1309107; Rho GTPase activating protein 11A; rho GTPase-activating protein 11A; Rho-type GTPase-activating protein 11A
Gene Symbols ARHGAP11A
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP11A(Accession Q6P4F7), corresponding to amino acid residues Q957-H1007.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228482, 9824
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.