Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101841 Supplier Thermo Scientific Supplier No. PA5101841
Only null left
Add to Cart
Add to Cart

ARHGAP17 Polyclonal Antibody

ARHGAP17 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP17
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Arhgap17
Gene Accession No. Q3UIA2, Q68EM7, Q99N37
Gene Alias 5730403H17Rik; Arhgap17; MST066; MST110; MSTP038; MSTP066; MSTP110; Nadrin; Nadrin2; neuron-associated developmentally regulated protein; neuron-associated developmentally-regulated protein; neuron-specific GTPase activating protein; neuron-specific GTPase-activating protein; PP367; PP4534; Rho GTPase activating protein 17; rho GTPase-activating protein 17; RhoGAP interacting with CIP4 homologs 1; RhoGAP interacting with CIP4 homologs protein 1; Rho-type GTPase-activating protein 17; Rich1; RICH-1; RICH1B; WBP15; WW domain-binding protein 15
Gene Symbols Arhgap17
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGAP17(Accession Q68EM7), corresponding to amino acid residues G623-R673.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55114, 63994, 70497
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.