Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP17 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568403
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8403 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8403 Supplier Bethyl Laboratories, Inc Supplier No. A304689AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA (MIM 165390), RAC1 (MIM 602048), and CDC42 (MIM 116952).

Specifications

Antigen ARHGAP17
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ARHGAP17
Gene Accession No. Q68EM7
Gene Alias MST066, MST110, MSTP038, MSTP066, MSTP110, NADRIN, PP367, PP4534, RICH1, RICH1B, WBP15, RICH-1, RhoGAP interacting with CIP4 homologs 1, neuron-associated developmentally regulated protein, rho GTPase-activating protein 17, rho-type GTPase-activating protein 17, rhoGAP interacting with CIP4 homologs protein 1
Gene Symbols ARHGAP17
Host Species Rabbit
Immunogen Between 757 and 625
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55114
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.