Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGAP18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520893
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520893 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520893 Supplier Invitrogen™ Supplier No. PA520893
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is NIH-3T3 cell lysate. PA5-20893 can be used with blocking peptide PEP-1007.

ARHGAP18 is one member of the human RhoGAP family with approximately 80 RhoGAP proteins known to be encoded in the human genome. Rho proteins belong to the Ras superfamily that is composed of over 50 members divided into six families, including Ras, Sar, Rho, Ran, Rab and Arf. Rho GTPases are important regulators of the actin cytoskeleton and consequently influence the shape and migration of cells. ARHGAP18 is linked to Ras, and thus, to EGFR-mediated proliferation, migration and differentiation. ARHGAP18 is precisely contained within chromosome 6q22-24, which has been shown to be linked to schizophrenia, suggesting that ARHGEP18 may play a role in this condition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP18
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ARHGAP18
Gene Accession No. Q8K0Q5, Q8N392
Gene Alias 4833419J07Rik; AI314906; AI843501; Arhgap18; bA307O14.2; MacGAP; Rho GTPase activating protein 18; rho GTPase-activating protein 18; Rho-type GTPase-activating protein 18; SENEX
Gene Symbols ARHGAP18
Host Species Rabbit
Immunogen A 15 amino acid peptide near the carboxy terminus of human ARHGAP18.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293947, 73910, 93663
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.