Learn More
Invitrogen™ ARHGAP5 Monoclonal Antibody (OTI9H2)

Description
ARHGAP5 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ARHGAP5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI9H2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ARHGAP5 |
| Gene Accession No. | Q13017 |
| Gene Alias | ARHGAP5; GFI2; growth factor independent 2; LRRGT00098; p100 RasGAP-associated p105 protein; p105 RhoGAP; p190B; p190-B; p190BRhoGAP; Rho GTPase activating protein 5; rho GTPase-activating protein 5; Rho GTPase-activating protein 5-like protein; RHOGAP5; rho-type GTPase-activating protein 5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.