Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555139 Supplier Invitrogen™ Supplier No. PA555139
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQPNSLSSEE PPTSEDQVGR EDSALARWAA DPANTAWMEN PEEAIYDDVP RENSDSEPDE MIYDDVENGD EGGNSSLEYG WSSSEFESYE EQSDSECK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 59%.

This gene encodes a Rho guanine nucleotide exchange factor (GEF). Rho GEFs regulate the activity of small Rho GTPases by stimulating the exchange of guanine diphosphate (GDP) for guanine triphosphate (GTP) and may play a role in neural morphogenesis. Mutations in this gene are associated with slowed nerve conduction velocity (SNCV). Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF10
Gene Accession No. O15013
Gene Alias 6430549H08Rik; Arhgef10; GEF10; Kiaa0294; mKIAA0294; RGD1565043; Rho guanine nucleotide exchange factor (GEF) 10; Rho guanine nucleotide exchange factor 10; SNCV
Gene Symbols ARHGEF10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGEF10. Recombinant protein control fragment (Product #RP-93758).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9639
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.