Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599586 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599586 Supplier Invitrogen™ Supplier No. PA599586
  • $573.00
    $472.65 / Each of 1

    Save $100.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARHGEF19.

Guanine nucleotide exchange factors (GEFs) such as ARHGEF19 accelerate the GTPase activity of Rho GTPases (see RHOA, MIM 165390).[supplied by OMIM, Dec 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARHGEF19
Gene Accession No. Q8IW93
Gene Alias 6030432F23; 6430573B13Rik; Arhgef19; Ephexin-2; Rho guanine nucleotide exchange factor (GEF) 19; Rho guanine nucleotide exchange factor 19; weakly similar to Rho GEF; weakly similar to Rho GEF 5; Wgef
Gene Symbols ARHGEF19
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGEF19(Accession Q8IW93), corresponding to amino acid residues L80-H130.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128272
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.