Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ARHGEF5 Recombinant Protein

Catalog No. 89011183 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-011-183 10 μg
89-011-184 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-011-183 Supplier Abnova™ Supplier No. H00007984P0110
Add to Cart
Add to Cart

Human ARHGEF5 full-length ORF recombinant protein with GST-tag at N-terminal

Rho GTPases play a fundamental role in numerous cellular processes initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein may be involved in the control of cytoskeletal organization.

  • Theoretical MW (kDa): 82.83
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MGGFSRRCSKLINSSQLLYQEYSDVVLNKEIQSQQRLESLSETPGPSSPRQPRKALVSSESYLQRLSMASSGSLWQEIPVVRNSTVLLSMTHEDQKLQEVKFELIVSEASYLRSLNIAVDHFQLSTSLRATLSNQEHQWLFSRLQDVRDVSATFLSDLEENFENNIFSFQVCDVVLNHAPDFRRVYLPYV
TNQTYQERTFQSLMNSNSNFREVLEKLESDPVCQRLSLKSFLILPFQRITRLKLLLQNILKRTQPGSSEEAEATKAHHALEQLIRDCNNNVQSMRRTEELIYLSQKIEFECKIFPLISQSRWLVKSGELTALEFSASPGLRRKLNTRPVHLHLFNDCLLLSRPREGSRFLVFDHAPFSSIRGEKCEMKLHGPHKNLFRLFLRQNTQGAQAEFLFRTETQSEKLRWISALAMPREELDLLECYNSPQVQCLRAYKPRENDELALEKADVVMVTQQSSDGWLEGVRLSDGERGWFPVQQVEFISNPEVRAQNLKEAHRVKTAKLQLVEQQA

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH14555
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 7984
Molecular Weight (g/mol) 82.83
Name ARHGEF5 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DKFZp686N1969/GEF5/P60/TIM/TIM1
Common Name ARHGEF5
Gene Symbol ARHGEF5
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.