Learn More
Invitrogen™ ARID1B Polyclonal Antibody

Description
ARID1B / BAF250B Polyclonal Antibody detects endogenous levels of ARID1B / BAF250B protein.
Specifications
Specifications
| Antigen | ARID1B |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ARID1B |
| Gene Accession No. | Q8NFD5 |
| Gene Alias | 6A3-5; 8030481M12; 9330189K18Rik; AI836955; Ardi1b; ARID domain-containing protein 1B; ARID1B; AT rich interactive domain 1B (Swi1 like); AT rich interactive domain 1B (SWI1-like); AT rich interactive domain 1B (SWI-like); AT-rich interaction domain 1B; AT-rich interactive domain-containing protein 1B; B230217J03Rik; BAF250B; BRG1-associated factor 250b; BRG1-binding protein ELD/OSA1; BRG1-binding protein hELD/OSA1; BRIGHT; CSS1; DAN15; ELD (eyelid)/OSA protein; ELD/OSA1; hOsa2; KIAA1235; mKIAA1235; MRD12; Osa homolog 2; OSA2; P250R; Smcf1; smcf-1; transcription factor 1 |
| Gene Symbols | ARID1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.