Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARID2 Monoclonal Antibody (GT7311)
GREENER_CHOICE

Catalog No. PIMA527862 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527862 Supplier Invitrogen™ Supplier No. MA527862
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 293T, HeLa, MHCC-97H, MEF, MEF KO Predicted reactivity: Mouse (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ARID2 is a subunit of the PBAF chromatin-remodeling complex which facilitates ligand-dependent transcriptional activation by nuclear receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID2
Applications Western Blot
Classification Monoclonal
Clone GT7311
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative; pH 7
Gene ARID2
Gene Accession No. Q68CP9
Gene Alias 1700124K17Rik; 4432409D24Rik; arid 2; ARID domain-containing protein 2; ARID2; AT rich interactive domain 2 (ARID, RFX-like); AT rich interactive domain 2 (Arid-rfx like); AT-rich interaction domain 2; AT-rich interactive domain-containing protein 2; Baf200; BRG1-associated factor 200; KIAA1557; p200; RFX-like; Zinc finger protein with activation potential; zipzap; zipzap/p200
Gene Symbols ARID2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human ARID2. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196528, 77044
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.