Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARID5A Monoclonal Antibody (GT1683)
GREENER_CHOICE

Catalog No. PIMA527814
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA527814 100 μL
1 options

Catalog No. PIMA527814

Supplier: Invitrogen™ MA527814

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 3xFlag human ARID5A-transfected 293T, THP-1, THP-1 nuclear extract Predicted Reactivity: Bovine (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID5A
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GT1683
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative
Gene ARID5A
Gene Accession No. Q03989, Q3U108
Gene Alias ARID domain-containing protein 5A; Arid5a; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interaction domain 5A; AT-rich interactive domain-containing protein 5A; D430024K22Rik; hypothetical protein LOC524118; LOW QUALITY PROTEIN: AT-rich interactive domain-containing protein 5A; modulator recognition factor 1; modulator recognition factor I; MRF1; MRF-1; RFVG5814; RP11-363D14
Gene Symbols ARID5A
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human ARID5A. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10865, 214855
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.