Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aripiprazole

Catalog No. p-7077705
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457990010 1 g
AC457990050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457990010 Supplier Thermo Scientific Chemicals Supplier No. 457990010
Only null left
Add to Cart

CAS: 129722-12-9 | C23H27Cl2N3O2 | 448.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129722-12-9
Molecular Formula C23H27Cl2N3O2
Molecular Weight (g/mol) 448.39
InChI Key CEUORZQYGODEFX-UHFFFAOYSA-N
Synonym aripiprazole, abilify, abilitat, abilify discmelt, discmelt, aripiprazol, opc 31, aripiprazole usan, aripiprazolum, aripirazole
PubChem CID 60795
ChEBI CHEBI:31236
IUPAC Name 7-[4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES C1CC(=O)NC2=C1C=CC(=C2)OCCCCN3CCN(CC3)C4=C(C(=CC=C4)Cl)Cl

Specifications

Melting Point 149.0°C
Loss on Drying 0.3% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetic acid, insoluble in methanol and hexane
Formula Weight 448.39
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Aripiprazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.