Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Arium™ Advance RO/Comfort II Softener

Catalog No. 14557604 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium™ Advance EDI
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-557-604 Arium™ Advance EDI
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-557-604 Supplier Sartorius Supplier No. H2OCSO1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arium™ Advance RO/Comfort II Softener consists of high efficient ion exchange resin, for efficient calcium removal.

  • Softener consisting of high efficient ion exchange resin, for efficient calcium removal
  • For intended use with:
    • Arium™ Advance EDI
    • Arium™ Comfort II
  • Quantity: 1 Unit/Pk.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Arium™ Advance EDI
Type Softener
Format Cartridge
For Use With (Application) Lab Water
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.