Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius arium™ comfort II ASTM 1, 2 Combined Water purification Systems with UV Lamp
GSA_VAGREENER_CHOICE

Catalog No. 15555516 Shop All Sartorius Products
Change view
Click to view available options
Flow Rate:
10 L/hr.
5 L/hr.
Mounting:
Benchtop
Wall
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Mounting
15-555-516 10 L/hr. Wall
15-555-510 5 L/hr. Benchtop
15-555-512 10 L/hr. Benchtop
15-555-514 5 L/hr. Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-555-516 Supplier Sartorius Supplier No. H2OII2UVB
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.

  • Equipped with latest EDI technology and unique cartridge kit
  • Innovative iJust technology optimizes product water quality
  • Touch display with intuitive menu navigation
  • Compact design with SD card slot
  • Performs electrochemical deionization in third purification stage
  • Reliable and safe removal of contamination contained in feed water
  • arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
  • Available as bench-top system for every laboratory workstation and wall-mounted with integrated wall bracket

Benefits

  • Optimized, economic water consumption
  • Premium product-water quality at all times
  • Guaranteed longer life of downstream ultrapure water systems

To install arium™ comfort II as built-in unit for supplying Type II pure water at flow rates of 5L/hour, please order corresponding bench-top system and conversion kit H2O-ACK-D

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 10 L/hr.
Product Water Type Type 1 and 2
Remote Dispenser No
Resistivity 18.2 Mohm.cm
Total Organic Carbon ≤2 ppb
Feedwater Pressure 2.0 to 6.9 bar
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC Analysis, Photometry, Microbiological media and reagents, Histology, ELISA, RIA
Humidity Range 80%
Includes UV lamp
Mounting Wall
Particle Size <1mL
Purification Method Adsorption using spherical activated carbon, catalyst, reverse osmosis, electrochemical deionization, optional final particle and sterile filtration
Turbidity <1 NTU
Width (English) 17 in.
Width (Metric) 43.5 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 18.7 in.
Depth (Metric) 47.6 cm
Weight (English) 61.7 lb. (Empty); 79.3 lb. (Operating)
Weight (Metric) 28 kg (Empty); 36 kg (Operating)
Electrical Requirements 100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.)
Daily Output 120 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Water Purification System
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.