Learn More
Sartorius arium™ Conversion Kit
GREENER_CHOICE
Catalog No. 15555528 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium ProArium Comfort I
Catalog No. For Use With (Equipment)
15-555-528 Arium ProArium Comfort I
1 options
Catalog No. 15-555-528 Supplier Sartorius Supplier No. H2OACKD
Add to Cart
Add to Cart
The arium™ conversion kit enables you to easily install an arium™ comfort or an arium™ pro bench-top system as a built-in unit.
The arium™ conversion kit extended tubing and its display and dispensing unit let you optimally integrate this equipment into your laboratory environment. This built-in version saves space on and above your lab bench as the control unit along with the display and dispenser permits variable wall-mounting options to suit your particular requirements.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium ProArium Comfort I
Type Conversion Kit
For Use With (Application) Water Purification
Material PVDF
Width (Metric) 3.4 m