Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius arium™ mini Scientific Pack

Catalog No. 14559045 Shop All Sartorius Products
Change view
Click to view available options
Capacity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14-559-045 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-559-045 Supplier Sartorius Supplier No. H2OSPACK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with arium mini and arium mini plus Ultrapure Water Systems

  • Deionization cartridge featuring top-down-flow technology
  • High performance capacity thanks to efficient ion exchange resins
  • Fast and effective adsorption of impurities through high-grade activated carbon
  • Best ion removal delivers ultrapure water that exceeds the ASTM Type 1 standard
TRUSTED_SUSTAINABILITY

Specifications

Description Sartorius arium™ mini Ultrapure Water System Accessory, Scientific Kit
Capacity 1 L
Feedwater Source Pretreated
Material Polypropylene
Purification Method Ion Exchange, Activated Carbon
Resin Type Ion Exchange Resin, Activated Carbon
Filter Type Spherical, catalytic activated carbon Ultrapure mixed bed ion exchange resin in semiconductor quality
For Use With (Equipment) arium mini and arium mini plus Ultrapure Water Systems
Type Scientific Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.