Learn More
Sartorius arium™ RO Module (advance EDI and comfort II systems)
GREENER_CHOICE
Catalog No. 15555526 Shop All Sartorius Products
Change view
Click to view available options
Includes:
1 RO Module
2 RO Modules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
15-555-526 2 RO Modules
15-555-525 1 RO Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-555-526 Supplier Sartorius Supplier No. H2OCROH2
Add to Cart
Add to Cart
The RO Module consists of two independent membranes whose design guarantees easy installation and reliable operation. Each module contains a low-energy reverse osmosis membrane in a polypropylene housing connections for feed water, product water and waste water. 1- and 2-module sets available.
The RO Modules typically enable high recovery rates. This optimizes the water consumption while still retaining at least up to 98% of the ions. Thanks to the backflush with permeate, particles and salts are removed from the surface of the membrane. This results in a longer service life and lower system maintenance costs. In addition, this backflush function on restarting the system allows for the immediate dispensing of high quality water.
TRUSTED_SUSTAINABILITY
Specifications
Type Reverse Osmosis Modules
For Use With (Application) Water Purification
Includes 2 RO Modules
Material Polypropylene
Height (Metric) 30.8 cm
Diameter (Metric) 7.8 cm
Weight (Metric) 0.468 kg