Learn More
Sartorius™ Arium™ SterilePlus
Catalog No. 14557601 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium™ Pro, Mini, Comfort, Remote Dispenser, Smart Station
Catalog No. For Use With (Equipment)
14-557-601 Arium™ Pro, Mini, Comfort, Remote Dispenser, Smart Station
1 options
Catalog No. 14-557-601 Supplier Sartorius™ Supplier No. 5441307H4CE
Add to Cart
Edge
Add to Cart
Sterile final filter for sterile and particle-free water dispensing.
  • Consists of combined 0.45 μm and 0.2 μm polyethersulfone membrane
  • For intended use with: Arium™ Mini, Mini Essential, Mini Plus, Pro, Pro DI, Pro UV, Pro UF, Pro VF, Comfort I, Comfort II, Remote Dispenser, Smart Station
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium™ Pro, Mini, Comfort, Remote Dispenser, Smart Station
Material PES
Pore Size (Metric) 0.45 μm and 0.2 μm