Learn More
Description
- Extended tubing set used for Smart Station ultrapure and pure for uncomplicated extension of standard supplied tubing set
- Operating range can be easily increased from 2 to 4 m
Specifications
Specifications
| Color (Overall) | White |
| For Use With (Equipment) | Arium™ Smart Station Pure Water |
| Type | Tubing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.