Learn More
Sartorius Arium™ Tubing Extension Set
Catalog No. 14556155 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium Smart Station Ultrapure Water
Arium™ Smart Station Pure Water
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-556-155 Arium Smart Station Ultrapure Water
14-556-154 Arium™ Smart Station Pure Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-556-155 Supplier Sartorius Supplier No. H2OATESUP
Add to Cart
Add to Cart
Extension set (4 m) for Arium™ Smart Station, used for uncomplicated extension of connection between Arium™ Smart Station and connected water system
  • Extended tubing set used for Smart Station ultrapure and pure for uncomplicated extension of standard supplied tubing set
  • Operating range can be easily increased from 2 to 4 m
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium Smart Station Ultrapure Water
Type Tubing