Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL13B Recombinant Rabbit Monoclonal Antibody (HL2173)
GREENER_CHOICE

Catalog No. FEMA550419 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550419 Supplier Invitrogen™ Supplier No. MA550419
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

ARL13B Recombinant Monoclonal Antibody for Western Blot, ICC/IF

This gene encodes a member of the ADP-ribosylation factor-like family. The encoded protein is a small GTPase that contains both N-terminal and C-terminal guanine nucleotide-binding motifs. This protein is localized in the cilia and plays a role in cilia formation and in maintenance of cilia. Mutations in this gene are the cause of Joubert syndrome 8. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL13B
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2173
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ARL13B
Gene Accession No. Q3SXY8
Gene Alias A530097K21Rik; A930014M17Rik; ADP ribosylation factor like GTPase 13B; ADP-ribosylation factor like GTPase 13B; ADP-ribosylation factor-like 13B; ADP-ribosylation factor-like 2-like 1; ADP-ribosylation factor-like protein 13B; ADP-ribosylation factor-like protein 2-like 1; Arl13b; Arl2l1; ARL2-like protein 1; C530009C10Rik; hnn; JBTS8; Protein hennin
Gene Symbols ARL13B
Host Species Rabbit
Immunogen Recombinant fragmemt of human ARL13B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200894, 304037
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.