Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560479 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560479 Supplier Invitrogen™ Supplier No. PA560479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TQLSASCVQL LGLLSAEQLA EASVLILFNK IDLPCYMSTE EMKSLIRLPD IIACAKQNIT TAEISAREGT GLAGVLAWLQ ATHRAND Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL16
Gene Accession No. Q0P5N6
Gene Alias ADP ribosylation factor like GTPase 16; ADP-ribosylation factor like GTPase 16; ADP-ribosylation factor-like 16; ADP-ribosylation factor-like protein 16; ARL16
Gene Symbols ARL16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL16. Recombinant protein control fragment (Product #RP-100325).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.