Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ARL17A Recombinant Protein Antigen
SDP

Catalog No. NBP259794X Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259794X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259794X Supplier R&D Systems™ Supplier No. NBP259794PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ARL17A The ARL17A Recombinant Protein Antigen is derived from E. coli. The ARL17A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Product Line Novus
Purity >80% by SDS-PAGE and Coomassie blue staining
Common Name ARL17A Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4
For Use With (Application) AC
Gene Alias ADP Ribosylation Factor Like GTPase 17A, ADP-Ribosylation Factor 1 Pseudogene 2, ADP-Ribosylation Factor 7, ADP-Ribosylation Factor 7 Variant, ADP-Ribosylation Factor Like GTPase 17A ADP-Ribosylation Factor-Like 17 Pseudogene 1, ADP-Ribosylation Factor-Like 17-Like, ADP-ribosylation factor-like protein 17, ARF1P2, ARL17A ARL17B, ARL17P1
Gene Symbol ARL17A
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 0.1 mL
Regulatory Status RUO
Source E.coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-53784. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.