Learn More
Description
Specifications
Specifications
| Detectable Analytes | Reagin (Syphilis) |
| Format | 30-well |
| Type | RPR Test Card |
| For Use With (Application) | ASI RPR test kit |
| Includes | 18mm circle cards, thirty-well |
| Quantity | 25 Pack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
