Learn More
Honeywell™ Armor Dot Aramid Fiber Cut-Resistant Gloves
Catalog No. 19014392
Change view
Click to view available options
:
Size: Lady's
Catalog No.
19-014-392 Size: Lady's
1 options
This item is not returnable. View return policy
Catalog No. 19-014-392 Supplier Honeywell™ Supplier No. KVAD18L
This item is not returnable. View return policy
Offer abrasion resistance and gripping ability with dexterity and flexibility.
  • Aramid fiber material provides cut resistance
  • PVC dots on both sides
  • Color: yellow glove/brown PVC dots
TRUSTED_SUSTAINABILITY
Specifications
Coating Material PVC Dots
Color Yellow
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Medium
Gender Women's
Glove Cuff Color Yellow
Glove Cuff Style Knit