Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorTuff™ Nitrile Coated Jersey Gloves - Knit Wrist

Catalog No. 19148277 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-277 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-148-277 Supplier PIP™ Supplier No. 563151/XL
  • $108.00
    $54.65 / Dozen of 12

    Save $53.35

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $561.00
    $284.65 / Case of 6 DZ

    Save $276.35

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Armor™ Series gloves combine low-coating deposit weight with high abrasion resistance to provide the protection of heavy-duty coated gloves, without sacrificing flexibility or tactile sensitivity.

  • ActiFresh™ treated to reduce germs and odors
  • Protects from physical hazards such as cuts, punctures, snags, and abrasions
  • Cotton jersey liner with blue nitrile coating
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Resistant to grease and oils
  • Launderable

Specifications

Coated Area Palm, Fingers, Knuckles
Coating Material Nitrile
Color Blue, Natural
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size X-Large
For Use With (Application) General Handling, Assembly, Fabrication, and Maintenance
Glove Cuff Style Knit
Series 56-3151
Length (English) 10.8 in.
Length (Metric) 27.5 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.