Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARMS Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564016
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4016 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4016 Supplier Bethyl Laboratories, Inc Supplier No. A303002AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

A novel protein kinase D-interacting substrate of 220 kDa (Kidins220) is the first identified Protein kinase D (PKD), a serine/threonine kinase. Kidins220 is an integral membrane protein that is expressed in brain and neuroendocrine cells. Kidins220 is phsphorylated by PKD at serine 918 in vivo.

Specifications

Antigen ARMS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KIDINS220
Gene Accession No. Q9ULH0
Gene Alias ARMS, MGC163482
Gene Symbols KIDINS220
Host Species Rabbit
Immunogen Between 1400 and 1450
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57498
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.