Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550911 Supplier Invitrogen™ Supplier No. PA550911
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARMCX1 Polyclonal Antibody detects endogenous levels of ARMCX1 protein.

This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and two Armadillo (arm) repeats. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX2 and ALEX3, on the X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMX1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARMCX1
Gene Accession No. Q9P291
Gene Alias 3010033I09Rik; AD032; ALEX1; ARM protein lost in epithelial cancers on chromosome X 1; arm protein lost in epithelial cancers, X chromosome, 1; armadillo repeat containing, X-linked 1; armadillo repeat-containing X-linked protein 1; ARMCX1; GASP7; Protein ALEX1
Gene Symbols ARMCX1
Host Species Rabbit
Immunogen Synthesized peptide derived from the N-terminal region of human ARMCX1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51309
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.