Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599252
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599252 Supplier Invitrogen™ Supplier No. PA599252
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARMX3.

This gene encodes a member of the ALEX family of proteins which may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and a single Armadillo (arm) repeat. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members on the X chromosome. Three transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMX3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARMCX3
Gene Accession No. Q5XID7, Q8BHS6, Q9UH62
Gene Alias 1200004E24Rik; AI450003; ALEX3; ARM protein lost in epithelial cancers on chromosome X 3; arm protein lost in epithelial cancers, X chromosome, 3; armadillo repeat containing X-linked 3; armadillo repeat containing, X-linked 3; armadillo repeat-containing X-linked protein 3; armadillo repeat-containing X-linked protein 3-like protein; Armcx3; BM-017; dJ545K15.2; DKFZp781N1954; GASP6; KIAA0443; MGC12199; Protein ALEX3; UNQ2517/PRO6007
Gene Symbols ARMCX3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARMCX3(Accession Q9UH62), corresponding to amino acid residues Q33-G83.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367902, 51566, 71703
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.