Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ARNT/HIF-1 beta Antibody Pack
SDP

Catalog No. NB100982 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100982 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100982 Supplier R&D Systems™ Supplier No. NB100982

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Includes

  • 1.NB100-110: ARNT/HIF-1 beta Antibody
  • 2.NB100-124: ARNT/HIF-1 beta Antibody (H1β234)

A Novus product, part of R&D Systems. For Research applications

ARNT/HIF-1 beta specifically detects ARNT/HIF-1 beta in Bovine, Ferret, Human, Mouse, Rat, Sheep samples. It is validated for ChIP Assay, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen ARNT/HIF-1 beta
Applications ChIP Assay, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Chromatin Immunoprecipitation (ChIP)
Gene Alias ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, HIF-1 Beta, HIF1B, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, nuclear translocator
Gene Symbols ARNT
Host Species Mouse
Immunogen See individual datasheets.
Quantity 2 Vials
Research Discipline Cancer, Hypoxia
Gene ID (Entrez) 405
Test Specificity See individual datasheets.
Includes 1.NB100-110: ARNT/HIF-1 beta Antibody
2.NB100-124: ARNT/HIF-1 beta Antibody (H1β234)
Target Species Bovine, Ferret, Human, Mouse, Rat, Sheep
Content And Storage Store at 4C. Do not freeze.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.