Learn More
Aroclor 1016, Restek
SDP
Catalog No. 06719724 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
Concentration:
200 μg/mL
50 mg/kg
500 mg/kg
Concentration or Composition Notes:
In Isooctane
In Transformer Oil
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration Concentration or Composition Notes
06-719-724 1 mL 500 mg/kg In Transformer Oil
06-719-712 1 mL 200 μg/mL In Isooctane
06-719-723 1 mL 50 mg/kg In Transformer Oil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-719-724 Supplier Restek Corporation Supplier No. 32076
Specifications
Concentration or Composition Notes In Transformer Oil
Concentration 500 mg/kg
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 500mg/kg