Learn More
PREDICT HEALTH LCMS/MS methods which are used for Phenylalanine, tyrosine and tryptophan analysis.
Supplier:  PREDICT HEALTH BR140010
Encompass_Preferred
Intended Use: Monitoring of dietary treatment of PKU Disorder: PhenylketonuriaAnalytes: Phenylalanine, Tyrosine, TryptophanAnalysis Method: HPLC with UV detectorEvaluation: QuantitativeSample Type: Plasma and Serum
Catalog No. NC2891057
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.