Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arp2 Recombinant Rabbit Monoclonal Antibody (5H2L7)

Catalog No. 703394
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-339-4 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-339-4 Supplier Invitrogen™ Supplier No. 703394
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Bovine, Pig. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

ARP2 (actin-related protein 2) is an ATP-binding component of the ARP2/3 complex - a multiprotein complex that mediates actin polymerization upon stimulation by the nucleation-promoting factor (NPF). The ARP2/3 complex mediates the formation of branched actin networks in the cytoplasm and provides the force for cell motility. The ARP2/3 complex also promotes actin polymerization in the nucleus, thereby regulating gene transcription and repair of damaged DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arp2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 5H2L7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene ACTR2
Gene Accession No. P61160, P61161
Gene Alias 4921510D23Rik; AA409782; Actin-like protein 14D; Actin-like protein 2; Actin-related protein 14 Da; Actin-related protein 14D; Actin-related protein 2; Actr14D; Actr2; Arp14D; ARP2; ARP2 actin related protein 2 homolog; ARP2 actin-related protein 2; ARP2 actin-related protein 2 homolog; ARP2 actin-related protein 2 homolog (yeast); Arp2-PA; Arp2-PB; Arp2-PC; CG9901; CG9901-PA; CG9901-PB; CG9901-PC; D6Ertd746e; Dmel\CG9901; Dmel_CG9901; I79_003843; LD18955p
Gene Symbols ACTR2
Host Species Rabbit
Immunogen Protein corresponding to human ARP2 [aa200-aa394].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10097, 66713
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.