Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arp3 Recombinant Rabbit Monoclonal Antibody (JB33-44)
GREENER_CHOICE

Catalog No. PIMA534721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534721 Supplier Invitrogen™ Supplier No. MA534721
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: A431, mouse placenta tissue lysate, mouse thymus tissue lysate, HUVEC, LOVO, SiHa, rat kidney tissue, human placenta tissue, human stomach cancer tissue, mouse kidney tissue, HL-60.

The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arp3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB33-44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ACTR3
Gene Accession No. P61158
Gene Alias 1200003A09Rik; Actin-like protein 3; actin-related protein 3; actin-related protein 3 homolog (yest); ACTR3; Arp3; ARP3 actin related protein 3 homolog; ARP3 actin-related protein 3; ARP3 actin-related protein 3 homolog; ARP3 actin-related protein 3 homolog (yeast)
Gene Symbols ACTR3
Host Species Rabbit
Immunogen Recombinant protein within Human Arp3 aa 308-418.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10096
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.