Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arrestin C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501726876 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6876 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6876 Supplier Proteintech Group Inc Supplier No. 111002AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Arrestin-C, also known as retinal cone Arrestin-3, X-Arrestin or cArr, is a member of the Arrestin family of proteins. It is predomitly found in the retina and pineal gland and localizes to the inner and outer segments of red-, green- and blue-cone photoreceptors and the inner plexiform regions. Two Arrestin-C isoforms exist due to alternative splicing. Isoform 1 is the mature full length protein and isoform 2 is truncated, ending with an arginine for amino acid residue 359. Arrestin-C expression is stimulated by retinoic acid. It may play a role in retina-specific signal transduction and bind to photoactivated-phosphorylated red/green opsins. In addition, Arrestin-C forms homodimers and oligomers with beta-Arrestins and may regulate beta-Arrestin mediated signaling.

Specifications

Antigen Arrestin C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ARR3
Gene Accession No. P36575, Q9EQP6
Gene Alias ARR3, Arrestin C, ARRX, C arrestin, CAR, cArr, Cone arrestin, Retinal cone arrestin 3, X arrestin
Gene Symbols ARR3
Host Species Rabbit
Immunogen Arrestin C Fusion Protein Ag1580
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170735, 407
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.