Learn More
Montana Molecular Borealis Arrestin PAR2 Assay - 10mL
SDP
Supplier:  Montana Molecular D2090G010
Encompass_Preferred
Measure arrestin signaling kinetics in real time with the live cell, fluorescent Borealis arrestin - PAR2 assay. Add the unmodified PAR2 receptor and Borealis sensor to cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. The kit includes the arrestin sensor BacMam, PAR2 BacMam, 500mM sodium butyrate in H2O, and a positive control receptor and agonist (AT1R BacMam and 5mM Angiotensin II in H2O). Borealis kits for several other receptors are also available, and larger-sized kits are available on request. Other Montana Molecular tools including the DAG and Calcium sensors can be used alongside this assay.
Catalog No. NC2600170
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.