Learn More
Apexbio Technology LLC ARRY-380 937265-83-3 5mg
SDP
Supplier:  Apexbio Technology LLC A83665
Encompass_Preferred
ARRY-380 (CAS 937265-83-3) is an orally bioavailable reversible ATP-competitive inhibitor of the HER2 (ErbB2) tyrosine kinase Demonstrating high selectivity approximately 500-fold greater for HER2 over EGFR ARRY-380 inhibits HER2 activity at nanomolar concentrations in vitro and is active against truncated HER2 isoforms such as p95-HER2 In preclinical models of ErbB2-driven intracranial tumors ARRY-380 improved survival and exhibited greater efficacy compared to other ErbB2-targeted agents Early clinical studies in patients with HER2-positive metastatic breast cancer reported tolerability without dose-limiting toxicity and evidence of antitumor activity ARRY-380 is a valuable tool for investigating HER2 signaling and evaluating targeted therapies in HER2-driven malignancies
Catalog No. 50-000-93832
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.