Learn More
Selleck Chemical LLC Bersacapavir-E1758-5MG
SDP
Supplier:  Selleck Chemical LLC E17585MG
Encompass_Preferred
Bersacapavir(JNJ-6379 JNJ-56136379) is a potent capsid assembly modulator (CAM) that accelerates capsid assembly and induces the formation of morphologically intact viral capsids devoid of genomic material It inhibits HBV replication with an EC50 of 54 nM in HepG2 117 cells
Catalog No. 50-313-9085
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.