Learn More
TARGETMOL CHEMICALS INC ARS-1323 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T103755MG
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. ARS-1323 is the racemate of ARS-1620 and a mutant K-ras G12C inhibitor. purity: 98%
Catalog No. 50-257-7458
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.