Learn More
Selleck Chemical LLC ARS-853
SDP
Supplier:  Selleck Chemical LLC S81561MG
Encompass_Preferred
ARS-853 is a selective covalent KRAS(G12C) inhibitor that inhibits mutant KRAS-driven signaling by binding to the GDP-bound oncoprotein and preventing activation ARS-853 also induces apoptosis
Catalog No. 50-136-5725
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.